Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056345 0.1 mg
NB10056345T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056345 Supplier Novus Biologicals Supplier No. NB10056345
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AKT3 Polyclonal specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen AKT3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9Y243
Gene Alias Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Gene Symbols AKT3
Host Species Rabbit
Immunogen This polyclonal antibody was raised against a synthetic peptide corresponding to amino acid residues 119-136 of human AKT3.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10000
Test Specificity A 50 kDa band is observed.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.