Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT3 Antibody (OTI9B2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27128A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27128A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27128A70 Supplier Novus Biologicals Supplier No. NBP271528AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKT3 Monoclonal specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AKT3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI9B2
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Gene Symbols AKT3
Host Species Mouse
Immunogen Full length recombinant protein of human AKT3 (NP_005456) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10000
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.