Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Akti-1/2
GREENER_CHOICE

Catalog No. p-7110904
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective dual Akt1 and 2 inhibitor

Akti-1/2 is a potent and selective dual Akt1 and 2 inhibitor (IC50 values are 50 and 210 nM, respectively). Selective for Akt1 and 2 over a panel of other tyrosine and serine/threonine kinases. Sensitizes LnCaP cells to TRAIL (TNF-related apoptosis-inducing ligand) induced apoptosis. Also enhances CAR and TCR retroviral transduction of human T cells. Active in vivo.

Specifications

Chemical Name or Material 1,3-Dihydro-1-[1-[[4-(6-phenyl-1 H-imidazo[4,5-g]quinoxalin-7-yl)phenyl]methyl]-4-piperidinyl]-2 H-benzimidazol-2-one
CAS 612847-09-3
Synonym Akt Inhibitor VIII
Quantity 50 mg
Target Akt (Protein Kinase B) Inhibitors
Molecular Formula C34H29N7O
Formula Weight 551.64
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.