Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AL 8697
Potent and selective p38α inhibitor
Specifications
Purity | >98% |
---|---|
Product Type | AL 8697 |
Molecular Weight (g/mol) | 402.41 |
Inhibitors | AL 8697 |
Description
Potent and selective p38α inhibitor (IC50 = 6 nM). Exhibits 14-fold greater inhibition of p38α compared to p38β; also displays 300-fold selectivity for p38α compared to a panel of 91 kinases. Exhibits anti-inflammatory properties.Specifications
>98% | |
AL 8697 | |
402.41 | |
AL 8697 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title