Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alas1 Antibody (2A9), Novus Biologicals™
SDP

Catalog No. NB297950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297950 50 μL
NB297951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297950 Supplier Novus Biologicals Supplier No. NBP32651350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Alas1 Monoclonal antibody specifically detects Alas1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Alas1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias 5-aminolevulinic acid synthase 1, ALAS, ALAS3, ALASH, ALAS-H, aminolevulinate, delta-, synthase 1, Delta-ALA synthase 1,5-aminolevulinate synthase, nonspecific, mitochondrial, Delta-aminolevulinate synthase 1, EC 2.3.1, EC 2.3.1.37, MIG4, migration-inducing protein 4
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Alas1 [UniProt P13196]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 211
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.