Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Antibody (188835) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB399927 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399927 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399927 Supplier Novus Biologicals Supplier No. FAB1455Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen Albumin
Applications CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 188835
Conjugate DyLight 680
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC
Formulation 50mM Sodium Borate
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Host Species Mouse
Immunogen Human Serum Albumin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 213
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.