Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB001764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001764 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001764 Supplier Novus Biologicals Supplier No. NB110163291000UL
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Albumin Polyclonal specifically detects Albumin in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Albumin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000-1:30000, ELISA 1:10000-1:50000, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 1:100-1:250, Immunohistochemistry-Paraffin 1:100-1:250
Formulation PBS and 0.2% BSA
Gene Accession No. P07724
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Goat
Immunogen Full length protein (Mouse).
Purification Method Affinity Purified
Quantity 1000 μL
Research Discipline Stem Cell Markers
Gene ID (Entrez) 213
Test Specificity By immunoelectrophoresis, the antiserum reacts specifically with mouse Albumin. No reactivity was detected against other mouse serum proteins.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.