Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Antibody, Novus Biologicals™
SDP

Catalog No. NB294842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294842 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294842 Supplier Novus Biologicals Supplier No. NBP325775
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Albumin Polyclonal antibody specifically detects Albumin in Human samples. It is validated for ELISA

Specifications

Antigen Albumin
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1/100 - 1/1000
Formulation PBS
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Host Species Sheep
Immunogen Native human serum albumin, 99% pure as assayed by agarose electrophoresis.
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213
Target Species Human
Content And Storage Store at -20 as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 for long term storage. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.