Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Sheep anti-Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Guinea Pig, Rabbit, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB018096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018096 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018096 Supplier Novus Biologicals Supplier No. NB12089400.25ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Sheep Polyclonal Antibody has been used in 3 publications

Albumin Polyclonal specifically detects Albumin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Radioimmunodiffusion.

Specifications

Antigen Albumin
Applications Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Immunohistochemistry
Classification Polyclonal
Concentration 19.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Radioimmunodiffusion
Formulation Glycine Buffered Saline (pH 7.4), 0.01% Benzamidine, 0.1% EACA and 1.0mM EDTA with 0.09% Sodium Azide
Gene Accession No. P02768
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Sheep
Immunogen Human serum albumin purified from Cohn Fraction V.
Purification Method IgG purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity This product gives a single arc when tested by IEP and 2D-IEP against human serum. Identity is confirmed by double diffusion (Ouchterlony) vs human plasma and a known anti-human albumin
Target Species Canine, Equine, Bovine, Rabbit, Human, Feline, Chicken, Rat, Guinea Pig, Pig, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.