Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALCAM/CD166 Antibody (2599C), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346926 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346926 Supplier Novus Biologicals Supplier No. FAB6563MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen ALCAM/CD166
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2599C
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166, Trp28-Ala526, Accession # Q13740
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 214
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.