Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALCAM/CD166 Antibody (311), DyLight 350
SDP

Catalog No. NB072435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072435 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072435 Supplier Novus Biologicals Supplier No. NBP290346UV

Rabbit Monoclonal Antibody

ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Mouse samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen ALCAM/CD166
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 311
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ALCAM/CD166 (NP_033785.1; Met1-Lys527).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 214
Test Specificity No cross-reactivity in ELISA with: Human ALCAM Human BCAM Human EpCAM Human CD146
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.