Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, Denatured, 200 Proof, Spectrum™ Chemical
SDP

Catalog No. 18600380 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Fiber Drum
Glass Bottle
Plastic Bottle
Steel Drum
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-380 6 x 1 L Glass Bottle
18-600-375 500 mL Glass Bottle
18-600-384 500 mL Plastic Bottle
18-600-377 1 L Glass Bottle
18-600-385 1 L Plastic Bottle
18-600-376 6 x 500 mL Glass Bottle
18-600-378 4 L Glass Bottle
18-600-379 4 L Plastic Bottle
18-600-386 6 x 1 L Plastic Bottle
18-600-381 4 x 4 L Glass Bottle
18-600-382 4 x 4 L Plastic Bottle
18-600-383 20 L Steel Drum
18-600-387 200 L Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 18-600-380 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A10381LTGLCS6
Add to Cart
Add to Cart

A1038, 64-17-5, C2H5OH

Spectrum™ Chemical Alcohol, Anhydrous, Denatured, 200 Proof is ethanol that has added additives that render it undrinkable, bad tasting or even poisonous to discourage people from consuming it. The additives do not chemically change the alcohol and thus it is identical to a normal alcohol apart from it's smell, color and taste and can be used for the exact same purposes as normal alcohol. Typical denaturants include methyl alcohol, isopropyl alcohol and methyl isobutyl ketone.

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0 , 108-10-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 85.8%, 4.3%, 8.96%, 0.91%
Quantity 6 x 1 L
Packaging Glass Bottle
UN Number UN1987
Percent Purity 84 to 88%
Grade Ungraded
Chemical Name or Material Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.