Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alcohol dehydrogenase 1A Antibody, Novus Biologicals™
SDP

Catalog No. NB396287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396287 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396287 Supplier Novus Biologicals Supplier No. NBP24686125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

alcohol dehydrogenase 1A Polyclonal antibody specifically detects alcohol dehydrogenase 1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen alcohol dehydrogenase 1A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2), 40% Glycerol
Gene Accession No. P07327
Gene Alias ADH, alpha subunit, ADH1alcohol dehydrogenase 1A, alcohol dehydrogenase 1 (class I), alpha polypeptide, alcohol dehydrogenase 1A (class I), alpha polypeptide, Alcohol dehydrogenase subunit alpha, aldehyde reductase, EC 1.1.1, EC 1.1.1.1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: IPQCGKCRICKNPESNYCLKNDVSNPQGTLQDGTSRFTCRRKPIHHF
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 124
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.