Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alcohol dehydrogenase 1B Antibody (OTI4F12), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270092APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270092APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270092APC Supplier Novus Biologicals Supplier No. NBP270092APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alcohol dehydrogenase 1B Monoclonal specifically detects alcohol dehydrogenase 1B in Human, Rat, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen alcohol dehydrogenase 1B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4F12
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADH, beta subunit, ADH2alcohol dehydrogenase 1B, alcohol dehydrogenase 1B (class I), beta polypeptide, alcohol dehydrogenase 2 (class I), beta polypeptide, Alcohol dehydrogenase subunit beta, aldehyde reductase, DKFZp686C06125, EC 1.1.1, EC 1.1.1.1
Gene Symbols ADH1B
Host Species Mouse
Immunogen Full length recombinant protein of human alcohol dehydrogenase 1B (NP_000659) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 125.0
Target Species Human, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.