Learn More
alcohol dehydrogenase 1B Antibody (OTI4F12), PerCP, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | alcohol dehydrogenase 1B |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI4F12 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | ADH, beta subunit, ADH2alcohol dehydrogenase 1B, alcohol dehydrogenase 1B (class I), beta polypeptide, alcohol dehydrogenase 2 (class I), beta polypeptide, Alcohol dehydrogenase subunit beta, aldehyde reductase, DKFZp686C06125, EC 1.1.1, EC 1.1.1.1 |
| Gene Symbols | ADH1B |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.