Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alcohol dehydrogenase 4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB126045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB126045 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB126045 Supplier Novus Biologicals Supplier No. NBP310572100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

alcohol dehydrogenase 4 Polyclonal specifically detects alcohol dehydrogenase 4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen alcohol dehydrogenase 4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias ADH-2, alcohol dehydrogenase 4, alcohol dehydrogenase 4 (class II), pi polypeptide, Alcohol dehydrogenase class II pi chain, aldehyde reductase, EC 1.1.1, EC 1.1.1.1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human alcohol dehydrogenase 4 (NP_000661). Peptide sequence PLTNLCGKISNLKSPASDQQLMEDKTSRFTCKGKPVYHFFGTSTFSQYTV
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 127
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.