Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ alcohol dehydrogenase 5 Protein
SDP

Catalog No. NBP19905310 Shop All R&D Systems Products
Click to view available options
Quantity:
100μg

Catalog No. NBP19905310

Supplier: Novus Biologicals™ NBP199053100UG

Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-374 of Human alcohol dehydrogenase 5 The Recombinant Human alcohol dehydrogenase 5 Protein is derived from E. coli. The Recombinant Human alcohol dehydrogenase 5 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH 7.5) containing 20% glycerol, 0.1M NaCl, 1mM DTT
Molecular Weight (g/mol) 42.3kDa
Purification Method SDS-PAGE
Quantity 100μg
Source Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name alcohol dehydrogenase 5
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.