Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alcohol dehydrogenase 7 Antibody (OTI2H10), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270098APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270098APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270098APC Supplier Novus Biologicals Supplier No. NBP270098APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alcohol dehydrogenase 7 Monoclonal specifically detects alcohol dehydrogenase 7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen alcohol dehydrogenase 7
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2H10
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADH4, ADH-4, alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide, alcohol dehydrogenase class 4 mu/sigma chain, Alcohol dehydrogenase class IV mu/sigma chain, alcohol dehydrogenase VII, alcohol dehydrogenase-7, class IV sigma-1 alcohol dehydrogenase, class IV sigmasigma alcohol dehydrogenase, EC 1.1.1, EC 1.1.1.1, Gastric alcohol dehydrogenase, Retinol dehydrogenase
Gene Symbols ADH7
Host Species Mouse
Immunogen Full length recombinant protein of human alcohol dehydrogenase 7 (NP_000664) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 131.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.