Learn More
Novus Biologicals™ alcohol dehydrogenase Activity Assay Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Gene ID (Entrez) | 124 |
| Gene Symbol | ADH1A |
| Quantity | 96 Tests |
| Synonym | ADH, alpha subunit, ADH1alcohol dehydrogenase 1A, alcohol dehydrogenase 1 (class I), alpha polypeptide, alcohol dehydrogenase 1A (class I), alpha polypeptide, Alcohol dehydrogenase subunit alpha, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 |
| Research Category | Cell Biology, Diabetes Research, Lipid and Metabolism |
| Species | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.