Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alcohol dehydrogenase Rabbit anti-Human, Mouse, Rat, Clone: 1X3C0, Novus Biologicals™
SDP

Catalog No. NB166740 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166740 20 μg
NB166739 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166740 Supplier Novus Biologicals Supplier No. NBP31582220UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alcohol dehydrogenase Monoclonal antibody specifically detects alcohol dehydrogenase in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen alcohol dehydrogenase
Applications Western Blot
Classification Monoclonal
Clone 1X3C0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ADH, alpha subunit, ADH1alcohol dehydrogenase 1A, alcohol dehydrogenase 1 (class I), alpha polypeptide, alcohol dehydrogenase 1A (class I), alpha polypeptide, Alcohol dehydrogenase subunit alpha, aldehyde reductase, EC 1.1.1, EC 1.1.1.1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human alcohol dehydrogenase (P07327).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 124
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.