Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Alda 1

Catalog No. 400510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-051-0 10 mg
40-055-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-051-0 Supplier Tocris Bioscience™ Supplier No. 4005/10
This item is not returnable. View return policy

ALDH2 activator; cardioprotective

Activator of aldehyde dehydrogenase 2 (ALDH2). Increases activity of wild-type ALDH2*1 and variant ALDH2*2 (by ∽2-fold and 11-fold respectively). Capable of partly restoring mutant ALDH2*2 activity; protective against cardiac ischemia.

Chemical Identifiers

CAS 349438-38-6
Molecular Formula C15H11Cl2NO3

Specifications

Quantity 10 mg
Formula Weight 324.16
Purity >99%
Molecular Weight (g/mol) 324.157
Chemical Name or Material Alda 1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.