Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Antibody (023), Janelia Fluor™ 549
SDP

Catalog No. NB109960 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109960 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109960 Supplier Novus Biologicals Supplier No. NBP289893JF549

Rabbit Monoclonal Antibody

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Aldehyde Dehydrogenase 1-A1/ALDH1A1
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 23
Conjugate Janelia Fluor 549
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM Sodium Borate
Gene Alias acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 (NP_000680.2; Ser 2-Ser 501).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 216
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.