Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Rabbit anti-Human, Clone: ALDH1A1/7011R, Novus Biologicals™
SDP

Catalog No. NB177716 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177716 100 μg
NB177717 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177716 Supplier Novus Biologicals Supplier No. NBP313880100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal antibody specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Aldehyde Dehydrogenase 1-A1/ALDH1A1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ALDH1A1/7011R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P00352
Gene Alias acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1
Host Species Rabbit
Immunogen Recombinant fragment (around aa 315-434) of human Aldehyde Dehydrogenase 1-A1/ALDH1A1 protein (exact sequence is proprietary). (Uniprot: P00352 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 216
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.