Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldehyde Dehydrogenase 3-A1/ALDH3A1 Antibody (OTI1B6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270152APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270152APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270152APC Supplier Novus Biologicals Supplier No. NBP270152APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldehyde Dehydrogenase 3-A1/ALDH3A1 Monoclonal specifically detects Aldehyde Dehydrogenase 3-A1/ALDH3A1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Aldehyde Dehydrogenase 3-A1/ALDH3A1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1B6
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Aldehyde dehydrogenase 3, aldehyde dehydrogenase 3 family, member A1, Aldehyde dehydrogenase family 3 member A1, aldehyde dehydrogenase isozyme 3, aldehyde dehydrogenase type III, ALDH3aldehyde dehydrogenase, dimeric NADP-preferring, ALDHIII, EC 1.2.1, EC 1.2.1.5, MGC10406, stomach aldehyde dehydrogenase
Gene Symbols ALDH3A1
Host Species Mouse
Immunogen Full length recombinant protein of human Aldehyde Dehydrogenase 3-A1/ALDH3A1 (NP_000682) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 218.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.