Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1A3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB019244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019244 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019244 Supplier Novus Biologicals Supplier No. NBP215339JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALDH1A3 Polyclonal specifically detects ALDH1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen ALDH1A3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50mM Sodium Borate
Gene Alias aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH1A3. The exact sequence is proprietary.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 220
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.