Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1A3 Antibody (OTI4E8), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB278769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB278769 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB278769 Supplier Novus Biologicals Supplier No. NBP270154MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALDH1A3 Monoclonal antibody specifically detects ALDH1A3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen ALDH1A3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4E8
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-100 and 413-512 of human ALDH1A3 (NP_000648) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 220
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.