Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1L2 Antibody, Novus Biologicals™
SDP

Catalog No. NB392917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392917 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392917 Supplier Novus Biologicals Supplier No. NBP26875025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALDH1L2 Polyclonal antibody specifically detects ALDH1L2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ALDH1L2
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias aldehyde dehydrogenase 1 family, member L2, aldehyde dehydrogenase family 1 member L2, DKFZp686A16126, DKFZp686M064, EC 1.5.1.6, FLJ36769, FLJ38508, MGC119536, MGC119537,10-formyltetrahydrofolate dehydrogenase ALDH1L2, Mitochondrial 10-formyltetrahydrofolate dehydrogenase, mtFDHDKFZp686P14145, probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: KCGGLQLQNEDVYMATKFEGFIQKVVRKLRGEDQEVELVVDYISKEVNEIMVKMPYQCFINGQFTDADDGKT
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 160428
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.