Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH2 Antibody (2C10), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB162411 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162411 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162411 Supplier Novus Biologicals Supplier No. NBP259488JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALDH2 Monoclonal antibody specifically detects ALDH2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen ALDH2
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2C10
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Host Species Mouse
Immunogen Recombinant human ALDH2 (18-517aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 217
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.