Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH2 Antibody (OTI4H2), PE, Novus Biologicals™
SDP

Catalog No. NBP270151PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270151PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270151PE Supplier Novus Biologicals Supplier No. NBP270151PE

Mouse Monoclonal Antibody

ALDH2 Monoclonal specifically detects ALDH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen ALDH2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4H2
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Gene Symbols ALDH2
Host Species Mouse
Immunogen Full length recombinant protein of human ALDH2 (NP_000681) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 217.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.