Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH4A1 Antibody (002), Biotin, Novus Biologicals™
SDP

Catalog No. NB037756 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037756 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037756 Supplier Novus Biologicals Supplier No. NBP290152B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ALDH4A1 Monoclonal specifically detects ALDH4A1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ALDH4A1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias aldehyde dehydrogenase 4 family, member A1, Aldehyde dehydrogenase family 4 member A1, ALDH4DKFZp779M035, delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial, EC 1.5.1.12, mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase, P5C dehydrogenase, P5CD, P5CDh
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH4A1 (AAH07581.1; Lys25-Gln563).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8659
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.