Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH6A1 Antibody (6H9B8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252410 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252410 Supplier Novus Biologicals Supplier No. NBP252410
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALDH6A1 Monoclonal specifically detects ALDH6A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen ALDH6A1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H9B8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias aldehyde dehydrogenase 6 family, member A1, Aldehyde dehydrogenase family 6 member A1, EC 1.2.1.18, EC 1.2.1.27, malonate-semialdehyde dehydrogenase, Malonate-semialdehyde dehydrogenase [acylating], methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial, MGC40271, MMSADHA, MMSDHmitochondrial acylating methylmalonate-semialdehyde dehydrogenase
Gene Symbols ALDH6A1
Host Species Mouse
Immunogen Purified recombinant fragment of human ALDH6A1 (AA: 1-195) expressed in E. Coli.
Molecular Weight of Antigen 57.8 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4329
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.