Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH6A1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132381 Supplier Novus Biologicals Supplier No. NBP132381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALDH6A1 Polyclonal specifically detects ALDH6A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ALDH6A1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias aldehyde dehydrogenase 6 family, member A1, Aldehyde dehydrogenase family 6 member A1, EC 1.2.1.18, EC 1.2.1.27, malonate-semialdehyde dehydrogenase, Malonate-semialdehyde dehydrogenase [acylating], methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial, MGC40271, MMSADHA, MMSDHmitochondrial acylating methylmalonate-semialdehyde dehydrogenase
Gene Symbols ALDH6A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH6A1. The exact sequence is proprietary.
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4329
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.