Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH7A1 Antibody (105), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB317263 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317263 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317263 Supplier Novus Biologicals Supplier No. NBP289927JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ALDH7A1 Monoclonal antibody specifically detects ALDH7A1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen ALDH7A1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 105
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH7A1 (Uniprot#: P49419-2; Ser 2-Gln 511).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 501
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.