Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH8A1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB325738 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325738 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325738 Supplier Novus Biologicals Supplier No. NBP299392APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALDH8A1 Polyclonal antibody specifically detects ALDH8A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ALDH8A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Aldehyde dehydrogenase 12, aldehyde dehydrogenase 8 family, member A1, aldehyde dehydrogenase family protein, ALDH12aldehyde dehydrogenase family 8 member A1, DJ352A20.2, DKFZp779D2315, EC 1.2.1, EC 1.2.1.-, EC 1.2.1.3, MGC138650
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ALDH8A1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64577
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.