Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH9A1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB058278 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB058278 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB058278 Supplier Novus Biologicals Supplier No. NBP299130AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALDH9A1 Polyclonal specifically detects ALDH9A1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ALDH9A1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias aldehyde dehydrogenase 9 family, member A1, Aldehyde dehydrogenase E3 isozyme, Aldehyde dehydrogenase family 9 member A1, ALDH4, ALDH7, ALDH9aldehyde dehydrogenase (NAD+), E34-trimethylaminobutyraldehyde dehydrogenase, EC 1.2.1, EC 1.2.1.19, EC 1.2.1.3, EC 1.2.1.47, EC 1.2.1.8, Gamma-aminobutyraldehyde dehydrogenase, R-aminobutyraldehyde dehydrogenase, TMABADH
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ALDH9A1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.