Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldo-keto Reductase 1C1/AKR1C1 Antibody (CPTC-AKR1C1-2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB379266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379266 Supplier Novus Biologicals Supplier No. NBP308655R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldo-keto Reductase 1C1/AKR1C1 Monoclonal antibody specifically detects Aldo-keto Reductase 1C1/AKR1C1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Aldo-keto Reductase 1C1/AKR1C1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-AKR1C1-2
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 20 alpha-hydroxysteroid dehydrogenase, 20-ALPHA-HSD, 20-alpha-hydroxysteroid dehydrogenase, aldo-keto reductase family 1 member C1, aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha(3-alpha)-hydroxysteroid dehydrogenase), C9, Chlordecone reductase homolog HAKRC, DD1/DD2, DD1MGC8954, DDHH-37, dihydrodiol dehydrogenase 1, Dihydrodiol dehydrogenase 1/2, dihydrodiol dehydrogenase isoform DD1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.112, EC 1.1.1.149,2-ALPHA-HSD, EC 1.3.1.20, HAKRCDDH1aldo-keto reductase C, HBAB, hepatic dihydrodiol dehydrogenase, High-affinity hepatic bile acid-binding protein, Indanol dehydrogenase, MBAB, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II 3-alpha-hydroxysteroid dehydrogenase
Host Species Mouse
Immunogen Recombinant human full-length Aldo-keto Reductase 1C1/AKR1C1 protein (Uniprot: Q04828)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1645
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.