Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldo-keto Reductase 1C4/AKR1C4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB045177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045177 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045177 Supplier Novus Biologicals Supplier No. NBP297581AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aldo-keto Reductase 1C4/AKR1C4 Polyclonal specifically detects Aldo-keto Reductase 1C4/AKR1C4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Aldo-keto Reductase 1C4/AKR1C4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 3-alpha-HSD, AKR1C4, aldo-keto reductase family 1, member C4, C11, CDR, CHDR, DD4, DD-4, HAKRA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Aldo-keto Reductase 1C4/AKR1C4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1109
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.