Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldolase A Antibody (3F9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP242620A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP242620A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP242620A30 Supplier Novus Biologicals Supplier No. NBP242620AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldolase A Monoclonal specifically detects Aldolase A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Aldolase A
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3F9
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALDA, aldolase A, fructose-bisphosphate, EC 4.1.2.13, fructose-1,6-bisphosphate triosephosphate-lyase, fructose-bisphosphate aldolase A, GSD12, Lung cancer antigen NY-LU-1, MGC10942, MGC17716, MGC17767, Muscle-type aldolase
Gene Symbols ALDOA
Host Species Mouse
Immunogen Recombinant human ALDOA (1-364aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 226.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.