Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldolase B Antibody (OTI3C6), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270162FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270162FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270162FR Supplier Novus Biologicals Supplier No. NBP270162FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldolase B Monoclonal specifically detects Aldolase B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Aldolase B
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3C6
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALDO2, aldolase B, fructose-bisphosphate, EC 4.1.2.13, fructose-bisphosphatase, fructose-bisphosphate aldolase B
Gene Symbols ALDOB
Host Species Mouse
Immunogen Full length recombinant protein of human Aldolase B (NP_000026) produced in HEK293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 229.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.