Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldolase C Antibody (1A1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP225144C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225144C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225144C Supplier Novus Biologicals Supplier No. NBP225144C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldolase C Monoclonal specifically detects Aldolase C in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Aldolase C
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1A1
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C
Gene Symbols ALDOC
Host Species Mouse
Immunogen The C-terminal 23 amino acids of human Aldolase C (KYEGSGEDGGAAAQSLYIANHAY). [UniProt# P09972]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 230
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.