Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alemtuzumab Antibody (2499A), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB315306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315306 Supplier Novus Biologicals Supplier No. FAB10246PECY55
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Alemtuzumab Monoclonal antibody specifically detects Alemtuzumab in Broad Species Reactivity samples. It is validated for Flow Cytometry

Specifications

Antigen Alemtuzumab
Applications Flow Cytometry
Classification Monoclonal
Clone 2499A
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Host Species Rabbit
Immunogen Full-length Alemtuzumab
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.