Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Mouse Anti-HCV Core Protein, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569919
Description
HCV Core Protein Monoclonal antibody specifically detects HCV Core Protein in Virus samples. It is validated for Intracellular staining (flow cytometry)
Specifications
HCV Core Protein | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Virus | |
Liquid |
Flow Cytometry | |
C7-50 | |
Alexa Fluor 647 | |
HCV Core Protein | |
Purified HCV Core Recombinant Protein | |
25 μg | |
951475 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction