Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Mouse Anti-HCV Core Protein, BD Pharmingen™
Mouse Monoclonal Antibody
$138.00 - $338.00
Specifications
Antigen | HCV Core Protein |
---|---|
Clone | C7-50 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
HCV Core Protein Monoclonal antibody specifically detects HCV Core Protein in Virus samples. It is validated for Intracellular staining (flow cytometry)Specifications
HCV Core Protein | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
951475 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
C7-50 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Virus | |
HCV Core Protein | |
Purified HCV Core Recombinant Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title