Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Mouse Anti-Human CD34, BD Pharmingen™
Mouse Monoclonal Antibody
$128.40 - $295.50
Specifications
| Antigen | CD34 |
|---|---|
| Clone | QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow cytometrySpecifications
| CD34 | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Human | |
| CD34; CD34 antigen | |
| Human Placental Vesicles | |
| Affinity Chromatography |
| QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| 947 | |
| IgG1 λ2 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title