Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG-2 Interacting Protein-X (ALIX0) Monoclonal Antibody (8E8), QED Bioscience™
SDP

Catalog No. 502278276
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8276 Supplier Leinco Technologies Inc Supplier No. 90001100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALG-2 Interacting Protein-X (ALIX0) Monoclonal antibody specifically detects ALG-2 Interacting Protein-X (ALIX0) in Human samples. It is validated for Immunohistochemistry, Western Blot

Specifications

Antigen ALG-2 Interacting Protein-X (ALIX0)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8E8
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris with 0.1M glycine, 2% sucrose and no preservative
Gene Pdcd6ip
Gene Accession No. Q8WUM4
Gene Alias AI480591; AIP1; ALG-2 interacting protein 1; ALG-2 interacting protein X; ALG-2-interacting protein 1; Alg2-interacting protein 1; ALG-2-interacting protein X; Alg2-interacting protein X; Alix; apoptosis-linked gene 2-interacting protein X; AW544830; C76364; dopamine receptor interacting protein 4; DRIP4; E2F1-inducible protein; Eig2; Hp95; KIAA1375; MGC17003; mKIAA1375; PDCD6-interacting protein; Pdcd6ip; pdcd6ip.L; programmed cell death 6 interacting protein; programmed cell death 6 interacting protein L homeolog; programmed cell death 6-interacting protein; putative signal tranduction protein Xp95; RGD1561176; Signal transduction protein Xp95; XELAEV_18031317mg; xp95
Host Species Mouse
Immunogen Recombinant human ALIX protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10015
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.