Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG13 Antibody, Novus Biologicals™
SDP

Catalog No. NB432687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432687 25 μL
NBP184330 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB432687 Supplier Novus Biologicals Supplier No. NBP18433025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALG13 Polyclonal specifically detects ALG13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ALG13
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9NP73
Gene Alias Asparagine-linked glycosylation 13 homolog, asparagine-linked glycosylation 13 homolog (S. cerevisiae), chromosome X open reading frame 45, CXorf45, EC 2.4.1.141, FLJ23018, FLJ31785, GLT28D1, glycosyltransferase 28 domain containing 1, Glycosyltransferase 28 domain-containing protein 1, hematopoietic stem/progenitor cells protein MDS031, MDS031, MGC12423, UDP-N-acetylglucosamine transferase subunit ALG13 homolog, YGL047W
Gene Symbols ALG13
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:VGTTSFDDLIACVSAPDSLQKIESLGYNRLILQIGRGTVVPEPFSTESFTLDVYRYKDSLKEDIQKADLVISHAGAGSCLETLEKGKPLVVVINEKLMNNHQLELAKQLHKEGHLFYCTCSTLPGLLQSMDLSTLKCYPPG
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79868
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.