Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG2 Antibody (OTI3C2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272308J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272308J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272308J549 Supplier Novus Biologicals Supplier No. NBP272308JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALG2 Monoclonal specifically detects ALG2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ALG2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C2
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alpha-1,3-mannosyltransferase ALG2, alpha-1,3-mannosyltransferase), asparagine-linked glycosylation 2 homolog (S. cerevisiae, asparagine-linked glycosylation 2 homolog (yeast, asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 2 homolog, CDGII, EC 2.4.1, EC 2.4.1.-, FLJ14511, GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase, hALPG2, homolog of yeast ALG2, NET38
Gene Symbols ALG2
Host Species Mouse
Immunogen Full length recombinant protein of human ALG2 (NP_149078) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 85365.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.