Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP247550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247550 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247550 Supplier Novus Biologicals Supplier No. NBP247550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALG8 Polyclonal specifically detects ALG8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ALG8
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias alpha-1,3-glucosyltransferase), asparagine-linked glycosylation 8 homolog (S. cerevisiae, asparagine-linked glycosylation 8 homolog (yeast, asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 8 homolog, CDG1H, dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase, Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase, dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase, EC 2.4.1, EC 2.4.1.-, HUSSY-02, MGC2840, probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase
Gene Symbols ALG8
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AVRECCKCIDGKKVGKELTEKPKF
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79053
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.