Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alginate Antibody (3G4-1F5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB259349700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259349700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259349700 Supplier Novus Biologicals Supplier No. NBP259349AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Alginate
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 3G4-1F5
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Gum levan, Kelacid, Levan gum, Sazio
Host Species Mouse
Immunogen Sodium Alginate conjugated to KLH
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.