Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alix Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB176474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176474 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176474 Supplier Novus Biologicals Supplier No. NBP149701AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Alix Polyclonal antibody specifically detects Alix in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Alix
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 818 and 868 of human Apoptosis-Linked Gene 2-Interacting Protein X using the numbering given in entry NP_037506.2 (GeneID 10015).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10015
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.